Learn More
Sino Biological Anti-Kallikrein 13 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10199T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human KLK13 / Kallikrein 13 (rh KLK13 / Kallikrein 13; Catalog#10199-H08H; NP_056411.1; Met1-Ile262). KLK13 / Kallikrein 13 specific IgG was purified by Human KLK13 / Kallikrein 13 affinity chromatography.
Catalog No. 50-172-2867