Learn More
Sino Biological Anti-CD57/B3gat1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101996T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Rat CD57, and purified by antigen affinity chromatography.
Catalog No. 50-172-2849