Learn More
Chemglass Life Sciences 102/75 Ball Joint, Clamp Size: 102, Tubing OD: 80mm
SDP
Catalog No. CG12015
Encompass_Preferred
102/75 Ball Joint, Clamp Size: 102, Tubing OD: 80mm. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. CG12015 Supplier Chemglass Life Sciences Supplier No. CG12015
102/75 Ball Joint, Clamp Size: 102, Tubing OD: 80mm. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.