Learn More
Chemglass Life Sciences 102/75 O-Ring Ball Joint, Clamp Size: 102, O-Ring Size: 238, Tubing OD: 80mm
SDP
Catalog No. CG12213
Encompass_Preferred
102/75 O-Ring Ball Joint, Clamp Size: 102, O-Ring Size: 238, Tubing OD: 80mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.
Catalog No. CG12213 Supplier Chemglass Life Sciences Supplier No. CG12213
102/75 O-Ring Ball Joint, Clamp Size: 102, O-Ring Size: 238, Tubing OD: 80mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.