Learn More
Ace Glass, Inc. 102/75 SOCKET JOINT (GLASSBLOWER)
SDP
Catalog No. 50979415
Encompass
Full length, standard wall 102/75 socket joint. This ACE socket joint is precision ground and vacuum leak-tested for fit and interchangeability.
Catalog No. 50-979-415 Supplier Ace Glass, Inc. Supplier No. 765578
Full length, standard wall 102/75 socket joint. This ACE socket joint is precision ground and vacuum leak-tested for fit and interchangeability.