Learn More
MSC 1/16" Id X 1/4" Od Dash 102 Silicone O-Ring
Catalog No. NC1466991
Encompass
3/32" Thick, Round Cross Section, Shore 65 - 75, -80 to 450°F
Catalog No. NC1466991 Supplier MSC Supplier No. 02047611
3/32" Thick, Round Cross Section, Shore 65 - 75, -80 to 450°F