Learn More
SOLVENTUM 1020 SCALE-UP CAP 05/60SP
Supplier:  SOLVENTUM 7000051627
Encompass_Preferred
Zeta Plus(TM) Encapsulated System Scale-Up Filter Capsule, EXT, with SP Series Media, 1 per case. Provide economical small scale clarification of pharmaceutical, biological, bioprocess, and cosmetic fluids. Our depth filtration media is composed of inorganic filter aid, cellulose and a positively charged resin system that that pulls negatively charged contaminants from the fluid.
Catalog No. NC0392536