Learn More
Sino Biological Anti-DC-SIGN/CD209 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10200MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD209 / DC-SIGN. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2895