Learn More
Sino Biological Anti-DC-SIGN/CD209 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10200MM01F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD209/DC-SIGN (rh CD209/DC-SIGN; Catalog#10200-H01H; NP_066978.1; Lys62-Ala404) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-2900