Learn More
Sino Biological Recombinant Anti-DC-SIGN/CD209 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10200R061100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human DC-SIGN / CD209 (rh DC-SIGN / CD209; Catalog#10200-H01H; NP_066978.1; Lys62-Ala404).
Catalog No. 50-172-2915