Learn More
Sino Biological Anti-OX40/TNFRSF4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102007T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Rat OX40/TNFRSF4/CD134, and purified by antigen affinity chromatography.
Catalog No. 50-172-2891