Learn More
Sino Biological Recombinant Human HER3/ERBB3 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10201H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Thr 643) of human ErbB3 (NP_001973.2) precursor was expressed with the C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-4300