Learn More
Sino Biological Recombinant Human HER3/ERBB3 Protein (aa 730-1065, His & GST Tag)
Supplier:  Sino Biological 10201H20B150
Encompass_Preferred
A DNA sequence encoding the cytoplasmic domain (Pro 730-Ser 1065) of human ErbB3 (NP_001973.2) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-4309