Learn More
Sino Biological Anti-HER3/ERBB3 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10201MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human HER3 / ErbB3 (rh HER3 / ErbB3; Catalog#10201-H08H; NP_001973.2; Met 1-Thr 643). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2949