Learn More
Sino Biological Anti-HER3/ERBB3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10201T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ERBB3/HER3 (rh ERBB3/HER3; Catalog#10201-H08H; NP_001973.2; Met1-Thr643). ERBB3/HER3 specific IgG was purified by Human ERBB3/HER3 affinity chromatography.
Catalog No. 50-172-2972