Learn More
Sino Biological Anti-MMP13 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102015T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MMP-13, and purified by antigen affinity chromatography.
Catalog No. 50-172-2929