Learn More
Sino Biological Anti-Leptin Receptor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102018T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Rat Leptin Receptor / CD295, and purified by antigen affinity chromatography.
Catalog No. 50-172-2935