Learn More
Sino Biological Human cytomegalovirus (HCMV) Glycoprotein B / gB Antibody, Mouse MAb
Supplier:  Sino Biological 10202MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CMV gB (Catalog#10202-V08H1; AAA45920.1) extracellular domain (Met 1-Lys 700) linked with the cytoplasmic domain (Arg 777-Val 907). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2981