Learn More
Sino Biological Human cytomegalovirus (HCMV) Glycoprotein B / gB Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 10202RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CMV gB (Catalog#10202-V08H1; AAA45920.1) extracellular domain (Met 1-Lys 700) linked with the cytoplasmic domain (Arg 777-Val 907). Human cytomegalovirus gB specific IgG was purified by Human CMV gB affinity chromatography.
Catalog No. 50-172-2987