Learn More
Sino Biological Human cytomegalovirus (HCMV) Glycoprotein B / gB Protein (Fc Tag)
Supplier:  Sino Biological 10202V02H1100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Lys 700) linked with the cytoplasmic domain (Arg 777-Val 907) of human CMV gB (AAA45920.1, with furin cleavage site mutated from 'RTKR' to 'TTQT') precursor was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4312