Learn More
Sino Biological Anti-DKK-3 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10203MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Dkk3 (rhDkk3; Catalog#10203-H08H; Met1-Ile350; AAF02676.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2995