Learn More
Sino Biological Anti-EVI2B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102030T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse EVI2B / CD361, and purified by antigen affinity chromatography.
Catalog No. 50-172-2989