Learn More
Sino Biological Recombinant Human E-Cadherin Protein (ECD, His Tag)
Supplier:  Sino Biological 10204H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human E-Cad precusor (NP_004351.1) (Met 1-Ile 707) was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-4324