Learn More
Sino Biological Anti-E-Cadherin Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10204MM08P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cadherin-1 / CDH1 / CD324 / E-cadherin (rh Cadherin-1 / CDH1 / CD324 / E-cadherin; Catalog#10204-H08H; NP_004351.1; Met 1-Ile 707) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-3032