Learn More
Sino Biological Recombinant Anti-E-Cadherin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10204R032100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human E-cadherin / CDH1 / Cadherin-1 / E-Cad (rh E-cadherin / CDH1 / Cadherin-1 / E-Cad; Catalog#10204-H08H; NP_004351.1; Met1-Ile707).
Catalog No. 50-172-3036