Learn More
Sino Biological Anti-E-Cadherin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10204RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Cadherin-1 / CDH1 (rh Cadherin-1; Catalog#10204-H08H; NP_004351.1; Met 1-Ile 707). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-3044