Learn More
Sino Biological Anti-TLR9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102040T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse TLR9, and purified by antigen affinity chromatography.
Catalog No. 50-172-3011