Learn More
Sino Biological Anti-Aromatase/CYP19A1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102059T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CYP19A1, and purified by antigen affinity chromatography.
Catalog No. 50-172-3074