Learn More
Sino Biological Recombinant Human ECE2 Protein (His Tag)
Supplier:  Sino Biological 10207H07H20
Encompass_Preferred
A DNA sequence encoding the ectodomain of human endothelin converting enzyme 2 isoform A (NP_055508.3) (Gly 199-Trp 883) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-4333