Learn More
Sino Biological Anti-PSMB8/LMP7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102075T46100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PSMB8, and purified by antigen affinity chromatography.
Catalog No. 50-172-3092