Learn More
Sino Biological Anti-eNOS Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102076T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Nos3 / eNOS, and purified by antigen affinity chromatography.
Catalog No. 50-172-3095