Learn More
Sino Biological Anti-Aquaporin 1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102077T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Aquaporin 1/AQP1, and purified by antigen affinity chromatography.
Catalog No. 50-172-3098