Learn More
Sino Biological Recombinant Human LAYN Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10208H08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Glu 220) of human Layilin (NP_849156.1) precursor was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4337