Learn More
Sino Biological Recombinant Human DDR2 Protein (hFc Tag)
Supplier:  Sino Biological 10209H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Arg 399) of human DDR2 precursor (NP_001014796.1) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4338