Learn More
Sino Biological Anti-Latexin Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10211MM02F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Latexin / Lxn (rh Latexin / Lxn; Catalog#10211-H07E; NP_064554.3; Glu 2-Glu 222) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-3163