Learn More
Sino Biological Anti-Latexin Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10211MM02P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Latexin (rh Latexin; Catalog#10211-H07E; NP_064554.3; Glu2-Glu222) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-3165