Learn More
Sino Biological Anti-Cathepsin K Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102123T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Cathepsin K, and purified by antigen affinity chromatography.
Catalog No. 50-172-3184