Learn More
Sino Biological Recombinant Human GFR Alpha-3/GFRA3 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10213H08H100
Encompass_Preferred
A DNA sequence encoding the human GFRa3 (NP_001487.2) (Met 1-Trp 382) was expressed, fused with the a C-terminal polyhistidine tag.
Catalog No. 50-161-4352