Learn More
Sino Biological Anti-GFR Alpha-3/GFRA3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10213RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human GFRA3 (rh GFRa3; Catalog#10213-H08H; NP_001487.2; Met 1-Trp 382). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-3216