Learn More
Sino Biological Recombinant Human BMP5 Protein (hFc Tag)
Supplier:  Sino Biological 10214H01H20
Encompass_Preferred
A DNA sequence encoding the carboxy-terminal domain (Gln 324-His 454) (the mature chain) of human BMP5 (NP_066551.1) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4355