Learn More
Sino Biological Recombinant Human Dectin-1/CLEC7A Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10215H01H100
Encompass_Preferred
A DNA sequence encoding the human Dectin1 (NP_072092.2) (Thr 66- Met201) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4356