Learn More
Sino Biological Anti-MVP Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102152T38100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human MVP/major vault protein, and purified by antigen affinity chromatography.
Catalog No. 50-172-3237