Learn More
Sino Biological Recombinant Human Fas/CD95/TNFRSF6 Protein (hFc Tag)
Supplier:  Sino Biological 10217H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Glu 173) of human Fas antigen (NP_000034.1) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4362