Learn More
Sino Biological Recombinant Human Fas/CD95/TNFRSF6 Protein (ECD, His Tag)
Supplier:  Sino Biological 10217H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Asn 173) of human FAS antigen (NP_000034.1) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-4364