Learn More
Sino Biological Anti-Fas/CD95/TNFRSF6 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10217MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD95 / APO-1 / TNFRSF6 (rh CD95 / APO-1 / TNFRSF6; Catalog#10217-H08H; NP_000034.1; Met1-Asn173). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3267