Learn More
Sino Biological Anti-Fas/CD95/TNFRSF6 Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10217MM10A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FAS / CD95 / APO-1 / TNFRSF6 (rh FAS / CD95 / APO-1 / TNFRSF6; Catalog#10217-H08H; NP_000034.1; Met1-Asn173) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-172-3271