Learn More
Sino Biological Anti-Fas/CD95/TNFRSF6 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10217MM10F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD95 / APO-1 / TNFRSF6 / FAS (rh CD95 / APO-1 / TNFRSF6 / FAS; Catalog#10217-H08H; NP_000034.1; Met 1-Asn 173) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-3273