Learn More
Sino Biological Anti-Fas/CD95/TNFRSF6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10217RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD95 (rh CD95; Catalog#10217-H08H; NP_000034.1; Met 1-Asn 173). CD95 specific IgG was purified by human CD95 affinity chromatography.
Catalog No. 50-172-3289