Learn More
Sino Biological Recombinant Human CSF3R/G-CSFR Protein (hFc Tag)
Supplier:  Sino Biological 10218H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Pro 621) of human G-CSF receptor (NP_000751.1) precursor was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4366