Learn More
Sino Biological Anti-CSF3R/G-CSFR Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10218MM06P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human G-CSFR / CD114 (rh G-CSFR / CD114; Catalog#10218-HCCH; NP_000751.1; Met 1-Pro 621) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-3312